4-Benzoylbutyric acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Oxo-5-phenylvaleric Acid, 5-Oxo-5-phenylpentanoic Acid, 5-Oxo-5-phenylpentanoic acid
L04013.06
L04013.14
L04013.06EA
1500
CZK
L04013.06
L04013.14
4-Benzoylbutyric acid 97%
4-Benzoylbutyric acid
Purity: 97%
CAS No.: 1501-05-9
Molecular Formula: C11H12O3
Formula Weight: 192.21
Melting Point: 126-128°
Beilstein Reference: 641947
MDL No.: MFCD00004411
Vzorec:
C₁₁H₁₂O₃ Bod tání: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004411 CAS číslo: 1501-05-9 EINECS: 216-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...