3-(Trifluoromethyl)crotonic acid 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L03816.06
L03816.14
L03816.06EA
2990
CZK
L03816.06
L03816.14
3-(Trifluoromethyl)crotonic acid 96%
3-(Trifluoromethyl)crotonic acid
Purity: 96%
CAS No.: 69056-67-3
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Flash Point: 79°(174°F)
Refractive Index: 1.3850
Melting Point: 28-30°
Boiling Point: 158-163°
Beilstein Reference: 1769204
UN No.: UN3261
MDL No.: MFCD00014374
Vzorec:
C₅H₅F₃O₂ MW: 154,09 g/mol Bod varu: 158…163 °C Bod tání: 28…30 °C Hustota: 1,318 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014374 CAS číslo: 69056-67-3 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...