Tisk…

Dimethyl-L-glutamate hydrochloride ≥97%

Dodavatel: Thermo Scientific

Synonyms: L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl

L03764.06 L03764.14
L03764.06EA 1420 CZK
L03764.06 L03764.14
Dimethyl-L-glutamate hydrochloride ≥97%
Dimethyl-L-glutamate hydrochloride

Purity: 98+% CAS No.: 23150-65-4 Molecular Formula: C7H13NO4.HCl Formula Weight: 211.65 Storage Temperature: 2°C to 8°C Odour: Odourless Melting Point: 99-103° Beilstein Reference: 4238829 Optical Rotation: +26° (c=5 in water) MDL No.: MFCD00038879

Vzorec: C₇H₁₃NO₄
MW: 211,64 g/mol
Bod tání: 99…103 °C
MDL Number: MFCD00038879
CAS číslo: 23150-65-4
EINECS: 245-461-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR