(1R)-endo-(+)-Fenchyl alcohol 96%
Dodavatel: Thermo Scientific
|
Varování
|
L03211.22
L03211.36
L03211.22EA
848
CZK
L03211.22
L03211.36
(1R)-endo-(+)-Fenchyl alcohol 96%
(1R)-endo-(+)-Fenchyl alcohol
Purity: 96%
CAS No.: 2217-02-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 73°(163°F)
Odour: Camphor-like
Melting Point: 37-44°
Boiling Point: 201-202°
Beilstein Reference: 1850917
Optical Rotation: +10.5° (c=10 in ethanol)
MDL No.: MFCD00003760
Vzorec:
C₁₀H₁₈O MW: 154,25 g/mol Bod varu: 201…202 °C Bod tání: 37…44 °C Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003760 CAS číslo: 2217-02-9 EINECS: 216-639-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...