3,5-Dimethyl-4-nitrobenzoic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
L02704.03
L02704.06
L02704.03EA
2330
CZK
L02704.03
L02704.06
3,5-Dimethyl-4-nitrobenzoic acid 97%
3,5-Dimethyl-4-nitrobenzoic acid
Purity: 97%
CAS No.: 3095-38-3
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Odour: Odourless
Melting Point: 222-226°
Beilstein Reference: 1965772
MDL No.: MFCD00014699
Vzorec:
C₉H₉NO₄ MW: 195,18 g/mol Bod tání: 222…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014699 CAS číslo: 3095-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...