Benzofuran-2-carboxaldehyde 99%
Dodavatel: Thermo Scientific
Synonyms:
Benzo[b]furan-2-carboxaldehyde, Benzofuran-2-carbaldehyde
L02456.04
L02456.09
L02456.04EA
3260
CZK
L02456.04
L02456.09
Benzofuran-2-carboxaldehyde 99%
Benzofuran-2-carboxaldehyde
Purity: 99%
CAS No.: 4265-16-1
Molecular Formula: C9H6O2
Formula Weight: 146.15
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6320
Boiling Point: 92-97°/1mm
Beilstein Reference: 2910
MDL No.: MFCD00015463
Vzorec:
C₉H₆O₂ MW: 146,145 g/mol Bod varu: 92…97 °C (1 mmHg) |
MDL Number:
MFCD00015463 CAS číslo: 4265-16-1 EINECS: 224-248-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...