Tisk…

1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride

L01833.06 L01833.22
L01833.06EA 869 CZK
L01833.06 L01833.22
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride

Purity: 98% CAS No.: 26487-67-2 Molecular Formula: C8H16ClN.HCl Formula Weight: 198.14 Melting Point: 206-212° Beilstein Reference: 3680388 UN No.: UN2811 MDL No.: MFCD00012842

Vzorec: C₈H₁₆ClN
MW: 198,13 g/mol
Bod tání: 206…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00012842
CAS číslo: 26487-67-2
EINECS: 247-733-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR