Tisk…

Cinnamyl bromide predominantly trans 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (3-Bromo-1-propen-1-yl)benzene, 3-Bromo-1-phenyl-1-propene

L01420.06 L01420.14 L01420.22
L01420.06EA 958 CZK
L01420.06 L01420.14 L01420.22
Cinnamyl bromide predominantly trans 95%
Cinnamyl bromide

Purity: 95% CAS No.: 4392-24-9 Molecular Formula: C6H5CH=CHCH2Br Formula Weight: 197.08 Flash Point: 87°(188°F) Refractive Index: 1.6210 Melting Point: 26-29° Boiling Point: 88-90°/1mm Beilstein Reference: 605833 UN No.: UN3261 MDL No.: MFCD00000245

Vzorec: C₉H₉Br
MW: 197.08 g/mol
Bod varu: 88…90 °C (1 mmHg)
Bod tání: 26…29 °C
Bod vzplanutí: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000245
CAS číslo: 4392-24-9
EINECS: 224-511-5
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR