2-(2-Pyridyl)benzimidazole ≥98%
Dodavatel: Thermo Scientific
Synonyms:
AI-4-88
L01339.03
L01339.06
L01339.03EA
704
CZK
L01339.03
L01339.06
2-(2-Pyridyl)benzimidazole ≥98%
2-(2-Pyridyl)benzimidazole
Purity: 98+%
CAS No.: 1137-68-4
Molecular Formula: C12H9N3
Formula Weight: 195.22
Melting Point: 221-225°
Beilstein Reference: 151411
MDL No.: MFCD00005586
Vzorec:
C₁₂H₉N₃ MW: 195,23 g/mol Bod tání: 221…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005586 CAS číslo: 1137-68-4 EINECS: 214-508-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...