Tisk…

Ethyl-5-hydroxyindole-2-carboxylate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 5-hydroxyindole-2-carboxylate

L00774.03 L00774.06
L00774.03EA 2690 CZK
L00774.03 L00774.06
Ethyl-5-hydroxyindole-2-carboxylate ≥98%
Ethyl-5-hydroxyindole-2-carboxylate

Purity: 98+% CAS No.: 24985-85-1 Molecular Formula: C11H11NO3 Formula Weight: 205.21 Storage Temperature: 2°C to 8°C Melting Point: 150-151° Beilstein Reference: 476925 MDL No.: MFCD00015458

Vzorec: C₁₁H₁₁NO₃
MW: 205,213 g/mol
Bod tání: 150…151 °C
MDL Number: MFCD00015458
CAS číslo: 24985-85-1
EINECS: 246-554-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR