Tisk…

Diethyl-2-bromo-2-methylmalonate 98%

Dodavatel: Thermo Scientific
Nebezpečí

L00575.14 L00575.22
L00575.14EA 2350 CZK
L00575.14 L00575.22
Diethyl-2-bromo-2-methylmalonate 98%
Diethyl-2-bromo-2-methylmalonate

Purity: 98% CAS No.: 29263-94-3 Molecular Formula: C8H13BrO4 Formula Weight: 253.10 Flash Point: 110°(230°F) Refractive Index: 1.4490 Boiling Point: 105-106°/5mm Beilstein Reference: 1779673 UN No.: UN3265 MDL No.: MFCD00009125

Vzorec: C₈H₁₃BrO₄
Bod varu: 105…106 °C (5 mmHg)
Hustota: 1,323
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009125
CAS číslo: 29263-94-3
EINECS: 249-541-6
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR