tert-Nitrosobutane dimer ≥98%
Dodavatel: Thermo Fisher Scientific
L00462.03
L00462.MD
L00462.03EA
2950
CZK
L00462.03
L00462.MD
tert-Nitrosobutane dimer ≥98%
tert-Nitrosobutane dimer
Purity: 98+%
CAS No.: 6841-96-9
Molecular Formula: C8H18N2O2
Formula Weight: 174.24
Storage Temperature: 2°C to 8°C
Melting Point: 88-92° subl.
MDL No.: MFCD00002065
Vzorec:
C₈H₁₈N₂O₂ Bod tání: 88…92 °C (subl.) |
MDL Number:
MFCD00002065 CAS číslo: 6841-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...