Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(E)-Ethyl 3-(pyrrolidin-1-yl)but-2-enoate
L00461.06
L00461.14
L00461.06EA
900
CZK
L00461.06
L00461.14
Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Ethyl (E)-3-(1-pyrrolidinyl)crotonate
Purity: 97%
CAS No.: 54716-02-8
Molecular Formula: C10H17NO2
Formula Weight: 183.25
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5410
Boiling Point: 125-126°/0.8mm
Beilstein Reference: 1310113
MDL No.: MFCD00014097
Vzorec:
C₁₀H₁₇NO₂ MW: 183,25 g/mol Bod varu: 125…126 °C (0,8 mmHg) |
MDL Number:
MFCD00014097 CAS číslo: 54716-02-8 EINECS: 259-303-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...