4-Nitrophenyl formate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L00316.06
L00316.14
L00316.06EA
1380
CZK
L00316.06
L00316.14
4-Nitrophenyl formate 98%
4-Nitrophenyl formate
Purity: 98%
CAS No.: 1865-01-6
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Storage Temperature: 2°C to 8°C
Melting Point: 70-74°
Beilstein Reference: 1950897
MDL No.: MFCD00014713
Vzorec:
C₇H₅NO₄ Bod tání: 70…74 °C |
MDL Number:
MFCD00014713 CAS číslo: 1865-01-6 EINECS: 217-473-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...