Tisk…

(Ethoxycarbonylmethyl)dimethylsulphonium bromide 98%

Dodavatel: Thermo Scientific
Varování

L00266.09 L00266.18
L00266.09EA 2300 CZK
L00266.09 L00266.18
(Ethoxycarbonylmethyl)dimethylsulphonium bromide 98%
(Ethoxycarbonylmethyl)dimethylsulphonium bromide

Purity: 98% CAS No.: 5187-82-6 Molecular Formula: CH3CH2O2CCH2S(CH3)2Br Formula Weight: 229.14 Storage Temperature: 2°C to 8°C Melting Point: ca 80° dec. Beilstein Reference: 4161472 MDL No.: MFCD00031604

Vzorec: CH₃CH₂O₂CCH₂S(CH₃)₂Br
Bod tání: ∼ 80 °C (rozkl.)
MDL Number: MFCD00830557
CAS číslo: 5187-82-6
EINECS: 225-968-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR