4-Phenylurazole ≥98%
Dodavatel: Thermo Fisher Scientific
L00255.06
L00255.14
L00255.06EA
1180
CZK
L00255.06
L00255.14
4-Phenylurazole ≥98%
4-Phenylurazole
Purity: 98+%
CAS No.: 15988-11-1
Molecular Formula: C8H7N3O2
Formula Weight: 177.16
Melting Point: 206-210°
Beilstein Reference: 163361
MDL No.: MFCD00005226
Vzorec:
C₈H₇N₃O₂ Bod tání: 206…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005226 CAS číslo: 15988-11-1 EINECS: 240-127-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...