Tisk…

Zlato 95%, nanoparticles 20 nm OD1 (Abs. 524 nm)

Dodavatel: Thermo Fisher Scientific

J67318.ACEA 3260 CZK
J67318.AC J67318.AE
Zlato 95%, nanoparticles 20 nm OD1 (Abs. 524 nm)
Zlato

Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red MDL No.: MFCD00003436


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Vzorec: Au
Storage Temperature: Refrigerator
MDL Number: MFCD00003436
CAS číslo: 7440-57-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR