Tisk…

Formamid ≥99.5%, Ultrapure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Carbamaldehyde

J67206.AE J67206.AK
J67206.AEEA 2120 CZK
J67206.AE J67206.AK
Formamid ≥99.5%, Ultrapure
Formamid

Purity: 99.5% min CAS No.: 75-12-7 Molecular Formula: CH3NO2 Formula Weight: 45.04 Flash Point: 150°(302°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Clear, Colourless Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 MDL No.: MFCD00007941

Vzorec: HCONH₂
MW: 45.04
Bod varu: 210 °C
Bod tání: 2 °C
Hustota: 1.134
Bod vzplanutí: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS číslo: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR