Tisk…

Bicine 99+% for electrophoresis

Dodavatel: Thermo Fisher Scientific

J67087.14 J67087.22
J67087.14EA 2950 CZK
J67087.14 J67087.22
Bicine 99+% for electrophoresis
Bicine

Purity: 99+% CAS No.: 150-25-4 Molecular Formula: C6H13NO4 Formula Weight: 163.17 Physical Form: Powder Appearance: White Melting Point: ca 191° dec. Beilstein Reference: 1769362 MDL No.: MFCD00004295

Vzorec: C₆H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS číslo: 150-25-4
EINECS: 205-755-1
Merck Index: 14,1201

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR