Tisk…

Ascomycin 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

J66751.MB J66751.MC
J66751.MBEA 7350 CZK
J66751.MB J66751.MC
Ascomycin 95%
Ascomycin

An analog of tacrolimus (FK-506). Ascomycin is an inhibitor of PP2 and it is potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases.


  • White


Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming. Very poorly soluble in water

Vzorec: C₄₃H₆₉NO₁₂
MW: 790 g/mol
Bod varu: 868 °C
Bod tání: 156…160 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Freezer
MDL Number: MFCD06198665
CAS číslo: 104987-12-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR