2-Merkaptoethanol 98+% for electrophoresis
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Hydroxyethyl mercaptan, β-Mercaptoethanol
J66742.0B
J66742.30
J66742.0BEA
3750
CZK
J66742.0B
J66742.30
2-Merkaptoethanol 98+% for electrophoresis
2-Merkaptoethanol
Purity: 98+%
CAS No.: 60-24-2
Molecular Formula: C2H6OS
Formula Weight: 78.13
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.114
Sensitivity: Air Sensitive; Hygroscopic
Appearance: Colourless
Odour: Unpleasant
Melting Point: -100°
Boiling Point: 156-158°
Beilstein Reference: 773648
UN No.: UN2966
MDL No.: MFCD00004890
Vzorec:
HSCH₂CH₂OH Bod varu: 156…158 °C Bod tání: –100 °C Hustota: 1,114 Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004890 CAS číslo: 60-24-2 EINECS: 200-464-6 UN: 2966 ADR: 6.1,II Merck Index: 14,5869 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...