Peroxodisíran draselný 99.0% for electrophoresis
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate
J66672.14
J66672.22
J66672.14EA
1380
CZK
J66672.14
J66672.22
Peroxodisíran draselný 99.0% for electrophoresis
Peroxodisíran draselný
Purity: 99.0%
CAS No.: 7727-21-1
Molecular Formula: K2S2O8
Formula Weight: 270.31
Physical Form: Crystalline
Density: 2.477
Appearance: White
Odour: Odourless
Melting Point: 100° dec.
UN No.: UN1492
MDL No.: MFCD00011386
Vzorec:
K₂S₂O₈ Hustota: 2,477 Storage Temperature: Ambient |
MDL Number:
MFCD00011386 CAS číslo: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: 5.1,III Merck Index: 14,7656 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...