Tisk…

Dimethylsulfoxid, Bioreagent

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

J66650.AD J66650.AE J66650.AK
J66650.ADEA 1900 CZK
J66650.AD J66650.AE J66650.AK
Dimethylsulfoxid, Bioreagent
Dimethylsulfoxid

CAS No.: 67-68-5 Molecular Formula: C2H6OS Formula Weight: 78.13 Flash Point: 87°(189°F) Physical Form: Liquid Density: 1.101 Refractive Index: 1.4790 Sensitivity: Hygroscopic Appearance: Clear, Colourless Odour: Mild Melting Point: 18.4° Boiling Point: 189° Beilstein Reference: 506008 MDL No.: MFCD00002089


Applications: Suitable for biotechnology applications such as peptide and oligonucleotide synthesis. This product is not sterile.

Vzorec: (CH₃)₂SO
MW: 78.14
Bod varu: 189 °C
Bod tání: 18.4 °C
Hustota: 1.101
Bod vzplanutí: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS číslo: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR