Pipemidic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylic acid
Purity: 98%
CAS No.: 51940-44-4
Molecular Formula: C14H17N5O3
Formula Weight: 303.32
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₁₄H₁₇N₅O₃ Bod varu: 717 °C Bod tání: 251…255 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00057291 CAS číslo: 51940-44-4 EINECS: 257-530-2 Merck Index: 14,7460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...