Mupirocin 92%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid
J66347.MC
J66347.MF
J66347.MCEA
22790
CZK
J66347.MC
J66347.MF
Mupirocin 92%
Mupirocin
Purity: 92%
CAS No.: 12650-69-0
Molecular Formula: C26H44O9
Formula Weight: 500.62
Flash Point:
Vzorec:
C₂₆H₄₄O₉ MW: 500,629 g/mol Bod varu: 672 °C Bod tání: 77…78 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01711620 CAS číslo: 12650-69-0 Merck Index: 14,6302 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...