Tisk…

Puromycin aminonucleoside 98%

Dodavatel: Thermo Fisher Scientific
Varování

J66301.MC J66301.ME
J66301.MCEA 21110 CZK
J66301.MC J66301.ME
Puromycin aminonucleoside 98%
Puromycin aminonucleoside

A puromycin aminonucleoside useful in the study of human glomerular disease.


  • White powder


Soluble in water at 50mg/ml. May require heating

Vzorec: C₁₂H₁₈N₆O₃
Bod tání: 215…216 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063462
CAS číslo: 58-60-6
EINECS: 200-388-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR