Tisk…

Sulbactam 98%

Dodavatel: Thermo Fisher Scientific
Varování

J66235.03 J66235.06
J66235.03EA 2680 CZK
J66235.03 J66235.06
Sulbactam 98%
Sulbactam

Purity: 98% CAS No.: 68373-14-8 Molecular Formula: C8H11NO5S Formula Weight: 233.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale brown Melting Point: 154-157° Solubility: Soluble in water at 18mg/ml MDL No.: MFCD00867005


Applications: Commonly used beta-lactam antibiotics to prevent degradation by beta-lactamase enzymes

Vzorec: C₈H₁₁NO₅S
Bod tání: 154…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00867005
CAS číslo: 68373-14-8
EINECS: 269-878-2
Merck Index: 14,8889

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR