Gatifloxacin sesquihydrate
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate
J66036.03
J66036.06
J66036.03EA
1330
CZK
J66036.03
J66036.06
Gatifloxacin sesquihydrate
Gatifloxacin sesquihydrate
CAS No.: 180200-66-2
Molecular Formula: C19H22FN3O4.1.5H2O
Formula Weight: 402.42
Physical Form: Powder
Appearance: White to pale yellow
Boiling Point: 684°
Solubility: Soluble in 1N NaOH at 10mg/ml
MDL No.: MFCD22373645
Applications: A broad spectrum antibiotic
Vzorec:
C₃₈H₅₀F₂N₆O₁₁ MW: 402,42 g/mol Bod varu: 684 °C Storage Temperature: Ambient |
MDL Number:
MFCD22373645 CAS číslo: 180200-66-2 Merck Index: 14,4376 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...