5-Bromo-3-indolyl-β-D-galactopyranoside ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside
Purity: 98+%
CAS No.: 97753-82-7
Molecular Formula: C14H16BrNO6
Formula Weight: 374.18
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00063691
Applications: alpha-Galactosidase substrate for Lac-gene detection systems
Vzorec:
C₁₄H₁₆BrNO₆ MW: 374,19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00063691 CAS číslo: 97753-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...