Leflunomide
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
J65917.MC
J65917.ME
J65917.MCEA
4360
CZK
J65917.MC
J65917.ME
Leflunomide
Leflunomide
CAS No.: 75706-12-6
Molecular Formula: C12H9F3N2O2
Formula Weight: 270.21
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Appearance: White
UN No.: UN2811
MDL No.: MFCD00867593
Applications: An immunosuppressive agent. Inhibits T and B cell proliferation.
Vzorec:
C₁₂H₉F₃N₂O₂ MW: 270,21 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00867593 CAS číslo: 75706-12-6 UN: 2811 ADR: 6.1,III Merck Index: 14,5432 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...