2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%
Dodavatel: Thermo Scientific
Synonyms:
Disodium 2'-deoxy-5'-O-phosphonatoguanosine, 2'-Deoxyguanosine-5'-monophosphoric acid disodium salt
J65186.03
J65186.06
J65186.14
J65186.03EA
1980
CZK
J65186.03
J65186.06
J65186.14
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate
Purity: 98%
CAS No.: 33430-61-4
Molecular Formula: C10H12N5Na2O7P.xH2O
Formula Weight: 391.18 (anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point:
Vzorec:
C₁₀H₁₂N₅Na₂O₇P·xH₂O MW: 391,18 g/mol (anhy) Bod tání: >245 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00080331 CAS číslo: 33430-61-4 EINECS: 251-517-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...