5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside 98%
Dodavatel: Thermo Scientific
Synonyms:
5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB
J64870.MF
J64870.MC
J64870.MD
J64870.MFEA
5430
CZK
J64870.MF
J64870.MC
J64870.MD
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside 98%
5,6-Dichlorobenzimidazole-1-beta-D-Ribofuranoside
Purity: 98%
CAS No.: 53-85-0
Molecular Formula: C12H12Cl2N2O4
Formula Weight: 319.14
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO to 75mM
Beilstein Reference: 39123
MDL No.: MFCD00036785
Applications: Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.
Vzorec:
C₁₂H₁₂Cl₂N₂O₄ MW: 319,14 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036785 CAS číslo: 53-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...