2'-Deoxyuridine-5'-triphosphate trisodium salt 95%
Dodavatel: Thermo Scientific
Synonyms:
dUTPsodium salt, Sodium ((2R,3S,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl triphosphate
J64716.MB
J64716.MC
J64716.MBEA
4120
CZK
J64716.MB
J64716.MC
2'-Deoxyuridine-5'-triphosphate trisodium salt 95%
2'-Deoxyuridine-5'-triphosphate trisodium salt
Purity: 95%
CAS No.: 102814-08-4
Molecular Formula: C9H15N2Na2O14P3
Formula Weight: 514.12
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00084701
Vzorec:
C₉H₁₅N₂Na₃O₁₄P₃ MW: 537,11 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00084701 CAS číslo: 102814-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...