Tisk…

2′-Deoxyuridine-5′-monophosphate disodium salt 99%

Dodavatel: Thermo Fisher Scientific
Varování

J64627.MD J64627.03
J64627.MDEA 3410 CZK
J64627.MD J64627.03
2′-Deoxyuridine-5′-monophosphate disodium salt 99%
2′-Deoxyuridine-5′-monophosphate disodium salt

Purity: 99% CAS No.: 42155-08-8 Molecular Formula: C9H11N2Na2O8P Formula Weight: 352.15 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00065282

Vzorec: C₉H₁₁N₂Na₂O₈P
Storage Temperature: Freezer
MDL Number: MFCD00065282
CAS číslo: 42155-08-8
EINECS: 255-687-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR