Adenosine-5'-diphosphate trilithium salt 98%
Dodavatel: Thermo Fisher Scientific
J64497.03
J64497.06
J64497.14
J64497.03EA
855
CZK
J64497.03
J64497.06
J64497.14
Adenosine-5'-diphosphate trilithium salt 98%
Adenosine-5'-diphosphate trilithium salt
Purity: 98%
CAS No.: 31008-64-7
Molecular Formula: C10H12Li3N5O10P2
Formula Weight: 445.00
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00065469
Vzorec:
C₁₀H₁₂Li₃N₅O₁₀P₂ Storage Temperature: Freezer |
MDL Number:
MFCD00065469 CAS číslo: 31008-64-7 EINECS: 250-431-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...