Tisk…

Locostatin

Dodavatel: Thermo Scientific

Synonyms: (S,E)-4-Benzyl-3-(but-2-enoyl)oxazolidin-2-one, UIC-1005, 4-Benzyl-3-[(2E)-but-2-enoyl]-1,3-oxazolidin-2-one, (4S)-N-Crotonyl-4-benzyl-2-oxazolidinone

J64425.MA J64425.MF
J64425.MAEA 4290 CZK
J64425.MA J64425.MF
Locostatin
Locostatin

CAS No.: 133812-16-5 Molecular Formula: C14H15NO3 Formula Weight: 245.27 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00278769


Applications: Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

Vzorec: C₁₄H₁₅NO₃
MW: 245,3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00278769
CAS číslo: 133812-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR