Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
ADIBO-NHS ester, with 4 carbon linker
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification.
- Moisture sensitive
It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
Soluble in DMF, THF, DMSO and DCM .
Upozornení: Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)