Tisk…

Amoxapine 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Chloro-11-(1-piperazinyl)dibenzo[b,f][1,4]oxazepine

J64066.MD J64066.03 J64066.06
J64066.MDEA 2110 CZK
J64066.MD J64066.03 J64066.06
Amoxapine 98%
Amoxapine

Purity: 98% CAS No.: 14028-44-5 Molecular Formula: C17H16ClN3O Formula Weight: 313.78 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in methanol, DMSO or chloroform. MDL No.: MFCD00069210


Applications: A tricyclic norepinephrine uptake inhibitor

Vzorec: C₁₇H₁₆ClN₃O
MW: 313,79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069210
CAS číslo: 14028-44-5
EINECS: 237-867-1
Merck Index: 14,576

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR