Isradipine ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Isopropyl Methyl 4-(4-Benzofurazanyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(4-Benzofurazanyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid Isopropyl Methyl Ester, Isopropyl Methyl 4-(2,1,3-Benzoxadiazol-4-yl)-1,4-dihydro-2,6-dim
Purity: 98+%
CAS No.: 75695-93-1
Molecular Formula: C19H21N3O5
Formula Weight: 371.39
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: Yellow
Solubility: Soluble in DMSO
MDL No.: MFCD00153820
Applications: L-type calcium channel blocker
Vzorec:
C₁₉H₂₁N₃O₅ MW: 371,39 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00153820 CAS číslo: 75695-93-1 Merck Index: 14,5240 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...