Tisk…

Amikacin disulphate

Dodavatel: Thermo Scientific

Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

J63862.14 J63862.22
J63862.14EA 5080 CZK
J63862.14 J63862.22
Amikacin disulphate
Amikacin disulphate

An aminoglycoside antibiotic derived from Kanamycin A.


Used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.


White to off-white powder


Soluble in water at 50 mg/ml with warming

Vzorec: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781,77 g/mol
Bod tání: 220…230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167475
CAS číslo: 39831-55-5
EINECS: 254-648-6
Merck Index: 14,405

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR