Harmol hydrochloride dihydrate 98%
Dodavatel: Thermo Scientific
J63827.03
J63827.06
J63827.03EA
2630
CZK
J63827.03
J63827.06
Harmol hydrochloride dihydrate 98%
Harmol hydrochloride dihydrate
Purity: 98%
CAS No.: 149022-16-2
Molecular Formula: C12H10N2O.HCl.2H2O
Formula Weight: 270.72 (234.69anhy)
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00150058
Applications: Induces apoptosis by Caspase-8 activation
Vzorec:
C₁₂H₁₀N₂O·HCl·2H₂O MW: 270,72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150058 CAS číslo: 149022-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...