Tisk…

Doxycycline monohydrate

Dodavatel: Thermo Fisher Scientific
Varování

J63805.06 J63805.14
J63805.06EA 2750 CZK
J63805.06 J63805.14
Doxycycline monohydrate
Doxycycline monohydrate

CAS No.: 17086-28-1 Molecular Formula: C22H24N2O8.H2O Formula Weight: 462.46 (444.44anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale yellow Melting Point: 167-168° Solubility: Soluble in ethanol or methanol. Sparingly soluble in water MDL No.: MFCD02682958


Applications: An antimicrobial. Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases

Vzorec: C₂₂H₂₄N₂O₈·H₂O
Bod tání: 167…168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682958
CAS číslo: 17086-28-1
Merck Index: 14,3440

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR