Sotalol hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
J63772.MC
J63772.03
J63772.06
J63772.MCEA
2670
CZK
J63772.MC
J63772.03
J63772.06
Sotalol hydrochloride 98%
Sotalol hydrochloride
Purity: 98%
CAS No.: 959-24-0
Molecular Formula: C12H20N2O3S.HCl
Formula Weight: 308.82
Physical Form: Crystalline powder
Appearance: White to off-white
MDL No.: MFCD00242937
Applications: Potent beta adrenergic antagonist
Vzorec:
C₁₂H₂₀N₂O₃S·HCl Storage Temperature: Ambient |
MDL Number:
MFCD00242937 CAS číslo: 959-24-0 EINECS: 213-496-0 Merck Index: 14,8728 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...