Tisk…

Ketotifen fumarate 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate

J63708.MD J63708.03
J63708.MDEA 1970 CZK
J63708.MD J63708.03
Ketotifen fumarate 99%
Ketotifen fumarate

Purity: 99% CAS No.: 34580-14-8 Molecular Formula: C19H19NOS.C4H4O4 Formula Weight: 425.50 Physical Form: Powder Appearance: White MDL No.: MFCD00079394


Applications: A H1-histamine receptor antagonist that inhibits the anaphylactic release of histamine

Vzorec: C₁₉H₁₉NOS
MW: 425.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079394
CAS číslo: 34580-14-8
EINECS: 252-100-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR