Tisk…

Clomiphene citrate salt ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

J63663.04 J63663.09
J63663.04EA 2350 CZK
J63663.04 J63663.09
Clomiphene citrate salt ≥98%
Clomiphene citrate salt

CAS No.: 50-41-9 Molecular Formula: C26H28ClNO.C6H8O7 Formula Weight: 598.08 Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00058322


Applications: A selective estrogen receptor modulator

Vzorec: C₃₂H₃₆ClNO₈
MW: 598.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058322
CAS číslo: 50-41-9
EINECS: 200-035-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR