Thiamphenicol
Dodavatel: Thermo Fisher Scientific
CAS No.: 15318-45-3
Molecular Formula: C12H15Cl2NO5S
Formula Weight: 356.22
Physical Form: Powder
Appearance: White to yellow
Solubility: Soluble in acetonitrile or DMF. Slightly soluble in water
Beilstein Reference: 2819542
MDL No.: MFCD00467983
Applications: The methyl-sulfonyl analogue of chloramphenicol
Vzorec:
C₁₂H₁₅Cl₂NO₅S Storage Temperature: Ambient |
MDL Number:
MFCD00467983 CAS číslo: 15318-45-3 EINECS: 239-355-3 Merck Index: 14,9301 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...