Tisk…

Zaprinast ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

J63326.MA J63326.MF
J63326.MAEA 1780 CZK
J63326.MA J63326.MF
Zaprinast ≥98%
Zaprinast

Purity: 98+% CAS No.: 37762-06-4 Molecular Formula: C13H13N5O2 Formula Weight: 271.28 Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00214073


Applications: Phosphodiesterase inhibitor

Vzorec: C₁₃H₁₃N₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00214073
CAS číslo: 37762-06-4
EINECS: 253-655-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR