Famotidine ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
J63165.03
J63165.06
J63165.03EA
1470
CZK
J63165.03
J63165.06
Famotidine ≥98%
Famotidine
Purity: 98+%
CAS No.: 76824-35-6
Molecular Formula: C8H15N7O2S3
Formula Weight: 337.45
Physical Form: Powder
Appearance: Off-white
Melting Point: 166-168°
Solubility: Soluble in water and methanol
MDL No.: MFCD00079297
Applications: A competitive histamine H2 receptor antagonist
Vzorec:
C₈H₁₅N₇O₂S₃ Bod tání: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079297 CAS číslo: 76824-35-6 Merck Index: 14,3930 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...