Tisk…

Rifamycin SV sodium ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione sodium salt, Rifamycin sodium

J63095.03 J63095.06
J63095.03EA 3090 CZK
J63095.03 J63095.06
Rifamycin SV sodium ≥98%
Rifamycin SV sodium

Purity: 98+% CAS No.: 14897-39-3 Molecular Formula: C37H46NNaO12 Formula Weight: 719.45 Physical Form: Powder Appearance: Orange to dark red MDL No.: MFCD00056847


Applications: Antibiotic effective against mycobacteria. Can also inhibit DNA polymerase

Vzorec: C₃₇H₄₆NNaO₁₂
MW: 719.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056847
CAS číslo: 14897-39-3
EINECS: 238-965-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR