Tobramycin sulphate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
J62995.MC
J62995.03
J62995.MCEA
2110
CZK
J62995.MC
J62995.03
Tobramycin sulphate
Tobramycin sulphate
CAS No.: 79645-27-5
Molecular Formula: (C18H37N5O9)2.5H2SO4
Formula Weight: 1425.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00133864
Applications: A water-soluble aminoglycoside antibiotic
Vzorec:
(C₁₈H₃₇N₅O₉)₂·5H₂SO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00133864 CAS číslo: 79645-27-5 Merck Index: 14,9490 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...